
2-Amino-4-methylthiophene-3-carboxamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4651-97-2
:
Formula:C₆H₈N₂OS
MDL Number:MFCD00052594
:156.20 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together