
3-Acetyl-5-bromopyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38940-62-4
:
Formula:C₇H₆BrNO
MDL Number:MFCD03086033
:200.03 g/mol
Melting Pt:77…79 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together