
2-Acetyl-6-bromopyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:49669-13-8
:
Formula:C₇H₆BrNO
MDL Number:MFCD00272200
:200.03 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C
Specification Test Results
Frequently Bought Together