
2-Acetyl-6-bromopyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:49669-13-8
:
Formula:C₇H₆BrNO
MDL Number:MFCD00272200
Melting Pt:51…55 °C
:200.03 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together