
5-Acetyl-2-bromopyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:139042-59-4
:
Formula:C₇H₆BrNO
MDL Number:MFCD04974527
:200.03 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together