Order Entry
Italy
ContactUsLinkComponent
2-(Methylthio)pyridine ≥98.0% (by GC)
Irritant
2-(Methylthio)pyridine ≥98.0% (by GC)
Catalog # TCIAM1849-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-(Methylthio)pyridine ≥98.0% (by GC)
Catalog # TCIAM1849-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:197°C
 :GHS07
CAS Number:18438-38-5
Formula:C₆H₇NS
MDL Number:MFCD01691593