Sodium hydrogen-4-amino-5-hydroxynaphthalene-2,7-disulphonate hydrate ≥80.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:343321-59-5
Formula:C₁₀H₈NNaO₇S₂·xH₂O
MDL Number:MFCD00036444
:341.28 g/mol (anhy)
EINECS:226-736-4
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (Neutralization titration):min. 80.0 %
- Purity (with Total Nitrogen):min. 80.0 %
- Drying loss:max. 6.0 %
- Sulfate:max. 5.0 %
Specification Test Results
Frequently Bought Together