
4,4'-Biphenyldiboronic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4151-80-8
:
Formula:C₁₂H₁₂B₂O₄
MDL Number:MFCD00151795
:241.85 g/mol
:GHS07
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together