

4-(Trifluoromethylsulphonyl)benzonitrile ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:312-21-0
:
Formula:C₈H₄F₃NO₂S
MDL Number:MFCD01631631
Melting Pt:87…92 °C
ADR:6.1,III
:235.18 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together