Order Entry
Italy
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Cotinine solution

Supelco® Organic Reference Standard, (−)-Cotinine solution

(−)-Cotinine solution, 1,0 mg/ml in methanol, Drug standard, 1 mL
Catalog # C0430-1ML
Supplier:   Merck
Supelco® Organic Reference Standard, (−)-Cotinine solution
Catalog # C0430-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, (−)-Cotinine solution
Supelco® Organic Reference Standard, (−)-Cotinine solution

(−)-Cotinine solution, 1,0 mg/ml in methanol, Drug standard, 1 mL

CAS Number:  
Price
86,70 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • GHS signal: GHS02, GHS06, GHS08

Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311

Specifications

Specifications

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Drug standard
  • Description:
    (−)-Cotinine solution
  • Cat. No.:
    C0430-1ML
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • MDL No.:
    MFCD00077696
  • Flash point:
    11 °C - Closed cup
  • Shipping conditions:
    STD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.