Order Entry
Italy
ContactUsLinkComponent
1-(Methylamino)-1-(methylthio)-2-nitroethene ≥99.0% (by HPLC)
Irritant
1-(Methylamino)-1-(methylthio)-2-nitroethene ≥99.0% (by HPLC)
Catalog # TCIAM1307-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1-(Methylamino)-1-(methylthio)-2-nitroethene ≥99.0% (by HPLC)
Catalog # TCIAM1307-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    110.0 to 115.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :148.18 g/mol
EINECS:263-266-9
 :GHS07
CAS Number:61832-41-5
Formula:C₄H₈N₂O₂S
MDL Number:MFCD00040994