Order Entry
Italy
ContactUsLinkComponent
N(α)-Benzyloxycarbonyl-L-lysine ≥97.0% (by HPLC, titration analysis)
N(α)-Benzyloxycarbonyl-L-lysine ≥97.0% (by HPLC, titration analysis)
  TCIAC1728-5G
 :  TCI
 :  
undefined
N(α)-Benzyloxycarbonyl-L-lysine ≥97.0% (by HPLC, titration analysis)
  TCIAC1728-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    -12.0 to -15.0 deg (C=2,0.2mol/L HCl)
  • Solubility in HCl (1+1):
    almost transparency

 

 

 : (S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH
 :
 :280.3 g/mol
 :218-662-6
 :2212-75-1
 :C₁₄H₂₀N₂O₄
 :MFCD00038204