
Ethyl-4-methylpyrrole-2-carboxylate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:40611-85-6
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00191668
:153.1800 g/mol
Boiling Pt:104°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:41.0 to 44.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together