
2-(1H-Pyrazol-3-yl)pyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75415-03-1
:
Formula:C₈H₇N₃
MDL Number:MFCD00115151
:145.16 g/mol
Melting Pt:121…122 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together