

4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19438-60-9
:
Formula:C₉H₁₂O₃
MDL Number:MFCD00005927
Flash Pt:164°C
:168.19 g/mol
EINECS:243-072-0
:GHS05
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Methanolysis Method):min. 97.0 %
Specification Test Results
Frequently Bought Together