Order Entry
Italy
ContactUsLinkComponent
1,1-Dimethylbiguanide hydrochloride, Millipore®
1,1-Dimethylbiguanide hydrochloride, Millipore®
Catalog # 317240-5
CAS Number:  
1,1-Dimethylbiguanide hydrochloride, Millipore®
Catalog # 317240-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :165.625 g/mol
EINECS:214-230-6
CAS Number:1115-70-4
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582