
5,6-Dimethylbenzotriazole ≥99.0% (by HPLC, titration analysis)
Catalog # TCIAD2177-5G
Supplier: TCI
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 % (calcd.on anh.substance)
- Melting point:152.0 to 157.0 deg-C
- Water:max. 15 %
Specification Test Results
About this item
:
:147.18 g/mol
EINECS:224-058-3
:GHS07
CAS Number:4184-79-6
Formula:C₈H₉N₃
MDL Number:MFCD00005703