
Sulpholane ≥80% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:126-33-0
:
Formula:C₄H₈O₂S
MDL Number:MFCD00005484
:120.17 g/mol
Merck Index:14,8955
Melting Pt:0…5 °C
Density:1.212
EINECS:204-783-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together