

1-Allylimidazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31410-01-2
:
Formula:C₆H₈N₂
MDL Number:MFCD00037894
:108.14 g/mol
Boiling Pt:212…213 °C
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:250-618-1
UN:3267
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow to light orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5030 to 1.5060 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together