N(α)-Acetyl-L-tryptophan ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1218-34-4
:
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00065976
:246.3 g/mol
Melting Pt:186 °C
Storage Temperature:Refrigerator
EINECS:214-935-9
Related Information
Frequently Bought Together