N(α)-Acetyl-D-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2280-01-5
:
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00065021
:246.3 g/mol
EINECS:218-912-4
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-24.2 to -25.9 deg (C=2.5, methanol)
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together