
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:171364-78-6
:
Formula:C₁₄H₂₂BNO₂
MDL Number:MFCD05663854
:247.1410 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together