
5,6-Dichloro-2-methylbenzimidazole ≥98.0% (by GC, titration analysis)
: TCI
:6478-79-1
:
:C₈H₆Cl₂N₂
:MFCD00276973
:201.0500 g/mol
:248°C
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together