



Methylamine 2 mol/l in THF, AcroSeal®, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-89-5
:
Formula:CH₅N
MDL Number:MFCD00008104
:31.06 g/mol
Density:0.861 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:200-820-0
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:1.8 to 2.2 M
Specification Test Results
Related Information
Frequently Bought Together