Order Entry
Italy
ContactUsLinkComponent
Hydrophobic interaction chromatography columns, ProPac™ HIC-10
Hydrophobic interaction chromatography columns, ProPac™ HIC-10
Certificates
Hydrophobic interaction chromatography columns, ProPac™ HIC-10
Specifications
  • Catalog No:
  • 555-7296
  • 555-7295
  • 555-7297
  • 555-7490
  • Endcapped:
  • N/A
  • N/A
  • N/A
  • N/A
  • Max. pressure:
  • 4000 psi
  • 4000 psi
  • 4000 psi
  • 4000 psi
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • Surface area:
  • 100 m²/g
  • 100 m²/g
  • 100 m²/g
  • 100 m²/g
  • Temperature limit:
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C