Safety shoes, Velcro® fastening, HACCP, Anatom, 2130/2135
Supplier: Abeba
Certificates
About this item
Anti-static safety shoes made from microfibre upper and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
- Perspiration-absorbent special lining with silver fibre
- Replaceable acc Wave insole; orthopaedic insoles possible
- Asymmetric heel and Flexzone for a natural bedding and foot movement
- TPU torsion waist, arch support and high stability
- Shock-absorbent PU midsole
- Sanitized® treatment reduces odour and bacterial growth
- Protective steel toecap (200 J)
- Washable at 30 °C
Suitable for use in kitchens.
Colour: White or black
Certifications: DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRC
Frequently Bought Together