Order Entry
Italy
ContactUsLinkComponent
Sodium thiocyanate ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Sodium thiocyanate ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3938-07
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Sodium thiocyanate ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3938-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 98.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • ACS - Assay (NaSCN):
    ≥ 98.0 %
  • ACS - Insoluble Matter:
    ≤ 0.005 %
  • ACS - Carbonate (as Na₂CO₃):
    ≤ 0.2 %
  • ACS - Chloride (Cl):
    ≤ 0.01 %
  • ACS - Sulfate (SO₄):
    ≤ 0.01 %
  • ACS - Sulfide (S):
    ≤ 0.001 %
  • ACS - Ammonium (NH₄):
    ≤ 0.002 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 2.0 ppm

Specification Test Results

About this item

 :
 :81.07 g/mol
Merck Index:13,09401
Melting Pt:287 °C
Density:1.735 g/cm³ (20 °C)
EINECS:208-754-4
 :GHS07
Storage Temperature:Ambient
CAS Number:540-72-7
Formula:NaSCN
MDL Number:MFCD00011123