Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vials
Powder vials, Chromacol™
Powder vials, Chromacol™
EPSC5.3SV-BC-GSK
:
EP SCIENTIFIC
Powder vials, Chromacol™
EPSC5.3SV-BC-GSK
:
EP SCIENTIFIC
Cap included:
No
Capacity:
5,00 ml
Colour:
Clear
Description:
Powder vial, with pre-applied barcode label to customer specification
Material:
Glass
Seq:
01
Pk:
187