



Dimethylamine ∼11% ∼2.0 mol/L in methanol
Catalog # TCIAD3292-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):11.0 to 13.0 w/w%
Specification Test Results
About this item
:
:45.08 g/mol
Merck Index:14,03228
Flash Pt:5°C
Storage Temperature:Ambient
EINECS:204-697-4
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:124-40-3
Formula:C₂H₇N
MDL Number:MFCD00008288