Order Entry
Italy
ContactUsLinkComponent
2-Bromohexane ≥70% (by GC) (contains 3-Bromohexane) stabilised
Flammable
2-Bromohexane ≥70% (by GC) (contains 3-Bromohexane) stabilised
Catalog # TCIAB1235-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
2-Bromohexane ≥70% (by GC) (contains 3-Bromohexane) stabilised
Catalog # TCIAB1235-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
 :165.07 g/mol
Boiling Pt:142 - 144 °C (1013 hPa)
Density:1,166 g/cm³ (20 °C)
Flash Pt:47°C
Storage Temperature:Ambient
EINECS:222-173-3
UN:1993
ADR:3,III
 :Stabilised with copper chip
 :GHS02
CAS Number:3377-86-4
Formula:H₃CBrCH(CH₂)₃CH₃
MDL Number:MFCD00039183