
Tetramethylammonium acetate ≥90%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10581-12-1
:
Formula:C₆H₁₅NO₂
MDL Number:MFCD00011630
:133.20 g/mol
Melting Pt:256 °C (dec.)
EINECS:234-179-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together