1-Hydroxy-2-naphthaldehyde ≥98.0% (by GC)
: TCI
:574-96-9
:
:C₁₁H₈O₂
:MFCD00092325
:172.1800 g/mol
:GHS07
- Environmentally Preferable:
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:53.0 to 57.0 deg-C
Frequently Bought Together