2-Ethoxy-1-naphthaldehyde ≥98.0% (by GC)
TCIAE0794-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:109.0 to 113.0 deg-C
:
:200.24 g/mol
:243-126-3
:GHS05
:19523-57-0
:C₁₃H₁₂O₂
:MFCD00046427