
Methyl-5-bromopicolinate ≥98%
Catalog # B25415.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:29682-15-3
:
Formula:C₇H₆BrNO₂
MDL Number:MFCD04112493
:216.03 g/mol
Melting Pt:99…103 °C
:GHS07
Storage Temperature:Ambient