Order Entry
Italy
ContactUsLinkComponent
Tetraethylammonium trifluoromethanesulphonate ≥99.0% (by titrimetric analysis)
Irritant
Tetraethylammonium trifluoromethanesulphonate ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Tetraethylammonium trifluoromethanesulphonate ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 99.0 %
  • Melting point:
    160.0 to 163.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together