Flammable 2,5-Dimethyl-3-hexanone ≥96.0% (by GC)
Catalog # TCIAD1453-5ML
CAS Number:

Flammable 2,5-Dimethyl-3-hexanone ≥96.0% (by GC)
Catalog # TCIAD1453-5ML
CAS Number:
Specification Test Results
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 96.0 %
Specification Test Results
About this item