Order Entry
Italy
ContactUsLinkComponent
 
1,4-Dibenzyloxybenzene ≥98.0% (by GC)
 :  TCI
 
undefined
1,4-Dibenzyloxybenzene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    127.0 to 131.0 deg-C
  • Solubility in hot Toluene:
    almost transparency

 

Frequently Bought Together