10-Chloro-3-decyne ≥97.0% (by GC)
Catalog # TCIAC1532-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:172.70 g/mol
Boiling Pt:122°C
CAS Number:18295-64-2
Formula:C₁₀H₁₇Cl
MDL Number:MFCD01321154