4-Bromo-4'-tert-butylbiphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:162258-89-1
:
Formula:C₁₆H₁₇Br
MDL Number:MFCD01321141
:289.21 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together