4-Bromo-4'-tert-butylbiphenyl ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:162258-89-1
:
Formula:C₁₆H₁₇Br
MDL Number:MFCD01321141
:289.21 g/mol
Boiling Pt:152 °C (2 mmHg)
Melting Pt:140…143 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together