

2,2,4-Trimethyl-1,3-dioxolane ≥98.0% (by GC)
Codice catalogo TCIAT2575-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:116.16 g/mol
Boiling Pt:98°C
Flash Pt:32°C
EINECS:214-766-0
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:1193-11-9
Formula:C₆H₁₂O₂
MDL Number:MFCD00090841