Order Entry
Italy
ContactUsLinkComponent
2-Mercaptothiophene ≥97.0% (by titrimetric analysis)
Toxic
2-Mercaptothiophene ≥97.0% (by titrimetric analysis)
Catalog # TCIAT1680-5G
Supplier:  TCI
CAS Number:  
undefined
Toxic
2-Mercaptothiophene ≥97.0% (by titrimetric analysis)
Catalog # TCIAT1680-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :116.20 g/mol
Boiling Pt:56°C
Flash Pt:65°C
Storage Temperature:Refrigerator
EINECS:231-881-1
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:7774-74-5
Formula:C₄H₄S₂
MDL Number:MFCD00051666