
2-Formyl-6-picoline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1122-72-1
:
Formula:C₇H₇NO
MDL Number:MFCD00006291
:121.14 g/mol
Flash Pt:68°C
EINECS:214-359-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together