Order Entry
Italy
ContactUsLinkComponent
2-Formyl-6-picoline ≥98%
Irritant
2-Formyl-6-picoline ≥98%
Catalog # H26841.14
CAS Number:  
Irritant
2-Formyl-6-picoline ≥98%
Catalog # H26841.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

CAS Number:1122-72-1
 :
Formula:C₇H₇NO
MDL Number:MFCD00006291
 :121.14 g/mol
Boiling Pt:105…108 °C (12 mmHg)
Melting Pt:29…33 °C
Flash Pt:68 °C (155 °F)
Storage Temperature:Ambient
EINECS:214-359-8
 :GHS07