1-Bromo-2-naphthaldehyde ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3378-82-3
:
Formula:C₁₁H₇BrO
MDL Number:MFCD00046368
:235.0767 g/mol
EINECS:222-180-1
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (GC):min. 96.0 %
- Melting point:115.0 to 120.0 deg-C
Specification Test Results
Frequently Bought Together