Order Entry
Italy
ContactUsLinkComponent
Acridine orange (hemizinc salt) 0.01% in aqueous solution
Acridine orange (hemizinc salt) 0.01% in aqueous solution
Catalog # CTEH62054/2
Supplier:  CLIN TECH
CAS Number:  
Acridine orange (hemizinc salt) 0.01% in aqueous solution
Catalog # CTEH62054/2
Supplier:  CLIN TECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :369.96 g/mol
EINECS:233-353-6
CAS Number:10127-02-3
Formula:C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MDL Number:MFCD00081043