Order Entry
Italy
ContactUsLinkComponent
Methylmagnesium bromide 35% ∼3 mol/L in ethyl ether
Flammable
Corrosive
Irritant
Health
Methylmagnesium bromide 35% ∼3 mol/L in ethyl ether
Catalog # TCIAM0785-250G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Methylmagnesium bromide 35% ∼3 mol/L in ethyl ether
Catalog # TCIAM0785-250G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    35.0 to 38.5 w/w%

Specification Test Results

About this item

 :
 :119.24 g/mol
Flash Pt:-40°C
EINECS:200-844-1
UN:1928
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:75-16-1
Formula:CH₃BrMg
MDL Number:MFCD00000041
Flash Pt:–40°C