Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Hydrogen peroxide 30%, Supelco®

 
 
 :   Merck
Hydrogen peroxide 30%, Supelco®
 :  Merck
 
Hydrogen peroxide 30%, Supelco®

 

 
   
 
  •  :7722-84-1
  •  :H₂O₂
  •  :MFCD00011333
  •  :34.01 g/mol
  •  :∼107 °C (1013 hPa)
  •  :< 0 °C
  •  :1.11 g/cm³ (20 °C)
  •  :Ambient
  •  :231-765-0
  •  :2014
  •  :5.1,II
  •  :Density: 1.11 g/cm³ (20 °C)

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 150,00 €
    150,00 €
    1 * 250 mL
       150,00 €

  • Plastic bottle
    1 L
       
       
    1 * 1 L
    1 * 1 L - 569,00 €
    569,00 €
    1 * 1 L
       569,00 €

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.