Order Entry
Italy
ContactUsLinkComponent
3,3,5-Trimethylcyclohexanol cis- and trans-mixture ≥90.0% (by GC)
Corrosive
3,3,5-Trimethylcyclohexanol cis- and trans-mixture ≥90.0% (by GC)
Catalog # TCIAT0472-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
3,3,5-Trimethylcyclohexanol cis- and trans-mixture ≥90.0% (by GC)
Catalog # TCIAT0472-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 90.0 % (total of isomer)

Specification Test Results

About this item

 :
 :142.24 g/mol
Boiling Pt:86°C
Melting Pt:30…32 °C
Density:0,9006 g/cm³ (16 °C)
Flash Pt:74°C
Storage Temperature:Ambient
EINECS:204-122-7
 :GHS05
CAS Number:116-02-9
Formula:C₉H₁₈O
MDL Number:MFCD00019378