
4-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6274-22-2
:
Formula:C₈H₁₀N₂O
MDL Number:MFCD00665792
:150.1778 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:178.0 to 184.0 deg-C
Specification Test Results
Frequently Bought Together